missing translation for 'onlineSavingsMsg'
Learn More

Ertapenem sodium, MedChemExpress

Product Code. 16465778
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465778 10 mg 10mg
16441937 50 mg 50mg
16416256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16465778 Supplier Medchem Express Supplier No. HY13625
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ertapenem sodium (L-749345) is a broad spectrum and long acting β-lactam antibiotic. Ertapenem sodium has a broad-spectrum anti-anaerobic activity against a variety of anaerobes with a mode MIC of 0.12 μg/mL. Ertapenem sodium can be used for the research of severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.

Ertapenem sodium (L-749345) is a broad spectrum and long acting β-lactam antibiotic. Ertapenem sodium has a broad-spectrum anti-anaerobic activity against a variety of anaerobes with a mode MIC of 0.12 μg/mL. Ertapenem sodium can be used for the research of severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.

Specifications

Chemical Name or Material Ertapenem sodium
Physical Form Powder
CAS 153773-82-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H24N3NaO7S
For Use With (Application) COVID-19-immunoregulation
Synonym L-749345 MK-826
Solubility Information H2O : 50 mg/mL (100.50 mM; Need ultrasonic)
SMILES O=C([C@H]1NC[C@@H](SC2=C(C(O)=O)N3[C@]([C@]([C@@H](C)O)([H])C3=O)([H])[C@H]2C)C1)NC4=CC=CC(C(O[Na])=O)=C4
Molecular Weight (g/mol) 497.5
Formula Weight 497.5
Percent Purity 98.86%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.