missing translation for 'onlineSavingsMsg'
Learn More

Melford Ertapenem Sodium Salt

Product Code. p-200087229
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018250

Brand: Melford E321001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.

Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.

It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria, including multidrug-resistant strains.

In the field of microbiology and infectious disease research, Ertapenem Sodium Salt is commonly used as a tool for studying bacterial susceptibility and resistance. It can be employed in antimicrobial susceptibility testing to determine the efficacy of different antibiotics against bacterial isolates.

Specifications

Chemical Name or Material Ertapenem Sodium Salt
CAS 153773-82-1
Quantity 1 g
Molecular Formula C22H24N3NaO7S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.