missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin A Enol Ether, TRC

Product Code. 17980865
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980865 10 mg 10mg
30009804 25 mg 25mg
17912225 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980865 Supplier Toronto Research Chemicals Supplier No. TRCE64997010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33396-29-1
Molecular Formula C37 H65 N O12
Molecular Weight (g/mol) 715.91
Synonym Erythromycin A Enol Ether, Erythromycin Imp. E (EP), Erythromycin, 8,9-didehydro-9-deoxo-6-deoxy-6,9-epoxy-, 6,15-Dioxabicyclo[10.2.1]pentadecane, erythromycin deriv., 8,9-Anhydroerythromycin A 6,9-hemiketal, BRL 46357ER, EM 201, Erythromycin A enol ether, ME 4, (2R,3R,4S,5R,8R,9S,10S,11R,12R)-9-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-5-ethyl-3,4dihydroxy-2,4,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-6,15dioxabicyclo[10.2.1]pentadec-1(14)-en-7-one
IUPAC Name (2R,3R,4S,5R,8R,9S,10S,11R,12R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-5-ethyl-3,4-dihydroxy-9-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-2,4,8,10,12,14-hexamethyl-6,15-dioxabicyclo[10.2.1]pentadec-1(14)-en-7-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@@]4(C)CC(=C(O4)[C@H](C)[C@@H](O)[C@]1(C)O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C37H65NO12/c1-14-25-37(10,43)30(40)20(4)28-18(2)16-36(9,50-28)32(49-34-27(39)24(38(11)12)15-19(3)45-34)21(5)29(22(6)33(42)47-25)48-26-17-35(8,44-13)31(41)23(7)46-26/h19-27,29-32,34,39-41,43H,14-17H2,1-13H3/t19-,20+,21+,22-,23+,24+,25-,26+,27-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
Formula Weight 715.4507
Percent Purity >95%
Chemical Name or Material Erythromycin A Enol Ether
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.