missing translation for 'onlineSavingsMsg'
Learn More

Erythropoietin/EPO Antibody (EPO/1367), HRP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54447H

Product Code. 18667236

  • 605.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Erythropoietin/EPO Monoclonal antibody specifically detects Erythropoietin/EPO in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Erythropoietin/EPO
Monoclonal
HRP
EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Recombinant fragment (around aa 28-162) of human Erythropoietin/EPO protein (exact sequence is proprietary) (Uniprot: P01588)
100 μg
Angiogenesis, Cancer, Cytokine Research, HIF Target Genes, Hypoxia
2056
Store at 4°C in the dark.
IgG1 κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
EPO/1367
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.