missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ESAM Monoclonal Antibody (340212)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561208
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
ESAM is a 55-kDa membrane protein composed of 2 extracellular Ig domains, a single transmembrane domain, and a 120 amino acid cytoplasmic domain. ESAM is related to members of the junctional adhesion molecule (JAM) family, and is expressed on endothelial cells and activated platelets. ESAM at endothelial tight junctions plays a role in the regulation of vascular permeability, in response to agents such as VEGF or extravasating neutrophils.
Specifications
| ESAM | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q925F2 | |
| ESAM | |
| Mouse myeloma cell line NS0-derived recombinant mouse ESAM, Gln30-Ser248 (predicted). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 340212 | |
| Unconjugated | |
| ESAM | |
| 2310008D05Rik; endothelial cell adhesion molecule; endothelial cell-selective adhesion molecule; endothelial cell-specific adhesion molecule; Esam; Esam1; HUEL (C4orf1)-interacting protein; LP4791 protein; UNQ220/PRO246; W117m | |
| Rat | |
| Protein A/G | |
| RUO | |
| 69524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction