missing translation for 'onlineSavingsMsg'
Learn More

Esculin, TRC

Product Code. 30180353
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180353 10 g 10g
30179765 5 g 5g
30180075 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180353 Supplier Toronto Research Chemicals Supplier No. TRCE65885010G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-75-9
Molecular Formula C15 H16 O9
Molecular Weight (g/mol) 340.28
Synonym 6-(β-D-Glucopyranosyloxy)-7-hydroxy-2H-1-benzopyran-2-one, Esculin, (-)-Esculin, 6,7-Dihydroxycoumarin 6-glucoside, 6-(β-D-Glucopyranosyloxy)-7-hydroxycoumarin, Aesculin, Crataegin, Enallachrome, Escosyl, Esculetin 6-O-glucoside, Esculetin 6-β-D-glucoside, Esculine, Esculoside, Polychrom, Polychrome, Vitamin C2
IUPAC Name 7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES OC[C@H]1O[C@@H](Oc2cc3C=CC(=O)Oc3cc2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H16O9/c16-5-10-12(19)13(20)14(21)15(24-10)23-9-3-6-1-2-11(18)22-8(6)4-7(9)17/h1-4,10,12-17,19-21H,5H2/t10-,12-,13+,14-,15-/m1/s1
Formula Weight 340.0794
Chemical Name or Material Esculin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.