missing translation for 'onlineSavingsMsg'
Learn More

Eslicarbazepine, MedChemExpress

Product Code. 16463256
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463256 25 mg 25mg
16437597 5 mg 5mg
16435636 100 mg 100mg
16424848 10 mg 10mg
16411107 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16463256 Supplier Medchem Express Supplier No. HY114703
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Eslicarbazepine is an oral anticonvulsant indicated for the adjunctive treatment of partial seizures.

Eslicarbazepine is an oral anticonvulsant indicated for the adjunctive treatment of partial seizures.

Specifications

Chemical Name or Material Eslicarbazepine
Physical Form Solid
CAS 104746-04-5
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C15H14N2O2
For Use With (Application) Neuroscience-Neurodegeneration
Synonym BIA 2-194
Solubility Information DMSO : 250 mg/mL (983.17 mM; Need ultrasonic)
SMILES O=C(N)N1C2=CC=CC=C2C[C@@H](C3=CC=CC=C31)O
Molecular Weight (g/mol) 254.28
Formula Weight 254.28
Percent Purity 99.43%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.