missing translation for 'onlineSavingsMsg'
Learn More

Estradiol 3-sulfamate, MedChemExpress

Product Code. 16482368
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482368 5 mg 5mg
16480686 20 mg 20mg
16480169 10 mg 10mg
16423159 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482368 Supplier Medchem Express Supplier No. HYU00112
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.

Estradiol 3-sulfamate (BLE 00084

Specifications

Chemical Name or Material Estradiol 3-sulfamate
CAS 172377-52-5
Color White
Purity Grade Notes Research
Molecular Formula C18H25NO4S
Quantity 5 mg
Synonym BLE 00084 E2MATE ES-J 995
Solubility Information DMSO : 62.5 mg/mL (177.83 mM; Need ultrasonic)
SMILES C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OS(N)(=O)=O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 351.46
Formula Weight 351.46
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.