missing translation for 'onlineSavingsMsg'
Learn More

Estradiol (cypionate), MedChemExpress

Product Code. 16492309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492309 Supplier Medchem Express Supplier No. HYB1100
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estradiol cypionate is a 17 β-cyclopentylpropinate ester of estradiol, inhibits ET-1 synthesis via estrogen receptor IC50 value: Target: estrogen receptor Estradiol cypionate is a synthetic ester, is a estrogen.

Estradiol cypionate is a 17 β-cyclopentylpropinate ester of estradiol, inhibits ET-1 synthesis via estrogen receptor

Specifications

Chemical Name or Material Estradiol (cypionate)
CAS 313-06-4
Color White
Purity Grade Notes Research
Molecular Formula C26H36O3
Quantity 1 g
Solubility Information DMSO : ≥ 30 mg/mL (75.65 mM)
SMILES C[C@@]12[C@@H](OC(CCC3CCCC3)=O)CC[C@@]1([H])[C@]4([H])CCC5=C(C=CC(O)=C5)[C@@]4([H])CC2
Molecular Weight (g/mol) 396.56
Formula Weight 396.56
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.