missing translation for 'onlineSavingsMsg'
Learn More

Estradiol valerianate, MedChemExpress

Product Code. 16441038
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441038 5 g 5g
16422259 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441038 Supplier Medchem Express Supplier No. HYB0672
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.

Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs.

Specifications

Chemical Name or Material Estradiol valerianate
CAS 979-32-8
Color White
Purity Grade Notes Research
Molecular Formula C23H32O3
Quantity 5 g
Synonym β-Estradiol 17-valerate
Solubility Information DMSO : ≥ 50 mg/mL (140.25 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]1([C@H]2OC(CCCC)=O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
Molecular Weight (g/mol) 356.5
Formula Weight 356.5
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.