missing translation for 'onlineSavingsMsg'
Learn More

Estramustine phosphate sodium, MedChemExpress

Product Code. 16475778
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475778 10 mg 10mg
16451937 50 mg 50mg
16426256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475778 Supplier Medchem Express Supplier No. HY13627
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.

Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.

Specifications

Chemical Name or Material Estramustine phosphate sodium
CAS 52205-73-9
Color White
Purity Grade Notes Research
Molecular Formula C23H30Cl2NNa2O6P
Quantity 10 mg
Solubility Information H2O : 62.5 mg/mL (110.75 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.86 mM; Need ultrasonic)
SMILES O=C(N(CCCl)CCCl)OC1=CC2=C(C=C1)[C@@]3([H])CC[C@@]4(C)[C@](CC[C@@H]4OP(O[Na])(O[Na])=O)([H])[C@]3([H])CC2
Molecular Weight (g/mol) 564.35
Formula Weight 564.35
Percent Purity 96.79%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.