missing translation for 'onlineSavingsMsg'
Learn More

Estrone, MedChemExpress

Product Code. 16478428
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478428 10 g 10g
16460838 5 g 5g
16412109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478428 Supplier Medchem Express Supplier No. HYB0234
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.

Estrone (E1) is a natural estrogenic hormone. Estrone is the main representative of the endogenous estrogens and is produced by several tissues, especially adipose tissue. Estrone is the result of the process of aromatization of androstenedione that occurs in fat cells.

Specifications

Chemical Name or Material Estrone
CAS 53-16-7
Color White
Purity Grade Notes Research
Molecular Formula C18H22O2
Quantity 10 g
Synonym E1 Oestrone
Solubility Information DMSO : 25 mg/mL (92.47 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.1 mg/mL (0.37 mM; Need ultrasonic)
SMILES C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O
Molecular Weight (g/mol) 270.37
Formula Weight 270.37
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.