missing translation for 'onlineSavingsMsg'
Learn More

Estropipate, MedChemExpress

Product Code. 16444587
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16444587 50 mg 50mg
16428698 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16444587 Supplier Medchem Express Supplier No. HYB1361
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.

Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.

Specifications

Chemical Name or Material Estropipate
CAS 7280-37-7
Color White
Purity Grade Notes Research
Molecular Formula C22H32N2O5S
Quantity 50 mg
Synonym Piperazine estrone sulfate Estrone sulfate piperazine salt
Solubility Information DMSO : 110 mg/mL (251.97 mM; Need ultrasonic)
SMILES C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(OS(=O)(O)=O)=C4)[C@@]3([H])CC1)C2=O.N5CCNCC5
Molecular Weight (g/mol) 436.56
Formula Weight 436.56
Percent Purity 99.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.