missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETAA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 572.00 €
Specifications
| Antigen | ETAA1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18151018
|
Novus Biologicals
NBP2-38229 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673756
|
Novus Biologicals
NBP2-38229-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ETAA1 Polyclonal specifically detects ETAA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ETAA1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9NY74 | |
| 54465 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: INNNSEHGAKNMFAISKQGSNLVQSKHLNPGSISVQTSLTNSSQIDKPMKMEKGEMYGNSPRFLGATNLTMYSKISNCQINNLHVSYTNTDVPIQVN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ETAA16, Ewing tumor-associated antigen 1, ewing's tumor-associated antigen 1, FLJ22647 | |
| ETAA1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title