missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ETAA1 Recombinant Rabbit Monoclonal Antibody (K01_1Q92)
GREENER_CHOICE

Product Code. 30826182
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826182 100 μL
30830404 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826182 Supplier Invitrogen™ Supplier No. MA563447

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ETAA1 Recombinant Monoclonal Antibody for Western Blot

ETAA1 (Ewing's tumor-associated antigen 1), also known as ETAA16, is a 926 amino acid cytoplasmic protein that is highly expressed in kidney, brain, liver and Ewing tumor cell lines. ETAA1 undergoes post-translational phosphorylation following DNA damage, most likely by either ATM or ATR, and is suggested to function as a tumour-specific cell surface antigen in Ewing's family of tumour cell lines. The gene encoding ETAA1 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETAA1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q92
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene ETAA1
Gene Accession No. Q9NY74
Gene Alias 5730466H23Rik; AA672646; Etaa1; ETAA16; Ewing tumor associated antigen 1; Ewing tumor-associated antigen 1; ewing's tumor-associated antigen 1; Ewing's tumor-associated antigen 1 homolog; ewing's tumor-associated antigen 16; RGD1560957
Gene Symbols ETAA1
Host Species Rabbit
Immunogen Recombinant protein of human ETAA1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54465
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.