Learn More
Invitrogen™ ETFA Monoclonal Antibody (24GB6605)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557397
 View more versions of this product
Product Code. 30275787
  • 198.00 € / 20µL
Description
ETFA Monoclonal Antibody for Western Blot
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ETFA
Monoclonal
Unconjugated
ETFA
2010200I21Rik; Alpha-ETF; D9Ertd394e; electron transfer flavoprotein alpha subunit; electron transfer flavoprotein alpha-subunit; electron transfer flavoprotein subunit alpha, mitochondrial; electron transfer flavoprotein, alpha polypeptide; electron transferring flavoprotein, alpha polypeptide; electron-transfer-flavoprotein alpha polypeptide; electron-transfer-flavoprotein, alpha polypeptide; EMA; ETF; ETFA; GA2; glutaric aciduria II; MADD
Mouse
Affinity chromatography
RUO
110842, 2108
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6605
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P13804, Q99LC5
ETFA
Recombinant protein of human ETFA (aa 1-333).
20 μL
Primary
Human, Mouse
Antibody
IgG1