missing translation for 'onlineSavingsMsg'
Learn More

Ethacridine lactate monohydrate, MedChemExpress

Product Code. 16458486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458486 Supplier Medchem Express Supplier No. HYB0889

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethacridine lactate (Acrinol) monohydrate is a widely used antiseptic and abortifacient. Ethacridine lactate monohydrate is effective against Staphylococcus aureus and other gram-positive cocci. Ethacridine lactate monohydrate is also a poly(ADP-ribose) glycohydrolase (PARG) inhibitor.

Ethacridine lactate (Acrinol) monohydrate is a widely used antiseptic and abortifacient. Ethacridine lactate monohydrate is effective against Staphylococcus aureus and other gram-positive cocci. Ethacridine lactate monohydrate is also a poly(ADP-ribose) glycohydrolase (PARG) inhibitor.

Specifications

Chemical Name or Material Ethacridine lactate monohydrate
Physical Form Solid
CAS 6402-23-9
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H23N3O5
For Use With (Application) COVID-19-immunoregulation
Synonym Acrinol monohydrate
Solubility Information DMSO : ≥ 100 mg/mL (276.71 mM) ∣H2O : 2 mg/mL (5.53 mM; Need ultrasonic)
SMILES CC(O)C(O)=O.NC1=CC2=NC3=CC=C(OCC)C=C3C(N)=C2C=C1.O
Molecular Weight (g/mol) 361.39
Formula Weight 361.39
Percent Purity 99.7%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.