missing translation for 'onlineSavingsMsg'
Learn More

Ethacrynic Acid, TRC

Product Code. 30138203
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138203 1 g 1g
17980415 100 mg 100mg
17901575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138203 Supplier Toronto Research Chemicals Supplier No. TRCE6760001G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-54-8
Molecular Formula C13 H12 Cl2 O4
Molecular Weight (g/mol) 303.14
Synonym Etacrynic acid, [2,3-Dichloro-4-(2-methylenebutanoyl)phenoxy]acetic acid, Acetic acid, 2-[2,3-dichloro-4-(2-methylene-1-oxobutyl)phenoxy], Acetic acid, [2,3-dichloro-4-(2-methylene-1-oxobutyl)phenoxy]- (9CI), Acetic acid, [2,3-dichloro-4-(2-methylenebutyryl)phenoxy]- (8CI), 2,3-Dichloro-4-(2-ethyl-1-oxo-2-propenyl)-phenoxyacetic acid, Crinuril, Edecril, Edecrin, Edecrina, Endecril, Etacrynic acid, Etakrinic acid, Ethacrynic acid, Hidromedin, Hydromedin, MK 595, Mingit, NSC 624008, NSC 85791, Otacril, Reomax, Taladren, Uregit, [2,3-Dichloro-4-(2-methylene-1-oxobutyl)phenoxy]acetic acid, [2,3-Dichloro-4-(2-methylenebutyryl)phenoxy]acetic acid, [4-(2-Methylenebutyryl)-2,3-dichlorophenoxy]acetic acid, [[2,3-Dichloro-4-[2-(methylene)butanoyl]phenyl]oxy]acetic acid, Etacrynic Acid
IUPAC Name 2-[2,3-dichloro-4-(2-methylidenebutanoyl)phenoxy]acetic acid
SMILES CCC(=C)C(=O)c1ccc(OCC(=O)O)c(Cl)c1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H12Cl2O4/c1-3-7(2)13(18)8-4-5-9(12(15)11(8)14)19-6-10(16)17/h4-5H,2-3,6H2,1H3,(H,16,17)
Formula Weight 302.0113
Percent Purity >95%
Chemical Name or Material Etacrynic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.