missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical
63.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
84.12 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
89.98 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
84.12 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
89.98 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Specifications
CAS | 64-17-5 |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Color | Colorless |
Molecular Formula | C2H6O |
MDL Number | 3568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
IUPAC Name | ethanol |
Molecular Weight (g/mol) | 46.069 |
Show More |
Safety and Handling
Product Identifier
- Ethanol (95% v/v)
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'