Learn More
Thermo Scientific Chemicals Ethidium bromide de-staining bags, with activation soln.
Description
Ethidium bromide de-staining bags is used to safely remove ethidium bromide from solution during overnight treatment. Each bag extracts up to 5 mg of Ethidium Bromide from solution thus minimizing the exposure of research personnel to this material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration. It provide a safer, more economical means for disposal of hazardous waste.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For removal of ethidium bromide from nucleotide solutions and gels
- Maximum capacity is 5mg ethidium bromide
- Also removes Coomassie stains
Specifications
Specifications
| Chemical Name or Material | Ethidium bromide de-staining bags |
| Name Note | with activation soln. |
| Quantity | 25 Ea. |
| Solubility Information | It is soluble in water. |
Frequently Asked Questions (FAQs)
Each ethidium bromide de-staining bag has a maximum capacity of 5 mg of ethidium bromide.
We recommend a one-time use for the de-staining bag for ease and convenience. Reusing the de-staining bag would require you to keep an accurate decontamination log to avoid saturating the resin.
We recommend following instructions for using the Ethidium bromide de-staining bags, with activation soln.:
- Wet the de-staining bag by placing it into approximately 5 mL of activation solution or water for 30 minutes.
- Add the wet bag to the solution containing ethidium bromide or stained gel.
- Allow the bag to mix overnight with gentle agitation or rocking. The gentle agitation increases the speed of adsorption.
- Remove the bag and dispose of as hazardous waste according to your institution's recommended procedures. Properly dispose of the gel or buffer solution.
Safety and Handling
- Ethidium bromide de-staining bag activation solution
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: propan-2-ol, Sodiumlauryl sulphate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.