missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide soln., 10mg/ml

Product Code. 15494679
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494679 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15494679 Supplier Thermo Scientific Alfa Aesar Supplier No. J62282.AB
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol

Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.1,2,3 It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA.4 Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Red
CAS 1239-45-8
Concentration 10mg/ml
Odor Odorless
Molecular Formula C21H20BrN3
MDL Number MFCD00011724
Quantity 10 mL
UN Number UN2810
Merck Index 14,4731
Physical Form Liquid
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
Solubility Information Soluble in water (10mg/mL).
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 5-ethyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
Molecular Weight (g/mol) 394.32
PubChem CID 14710
ChEBI CHEBI:4883
Formula Weight 394.32
Chemical Name or Material Ethidium bromide soln.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethidium bromide solution, 10 mg/ml
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 3,8-Diamino-1-ethyl-6-phenylphenantridinium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.