missing translation for 'onlineSavingsMsg'
Learn More

Ethionamide, MedChemExpress

Product Code. 16442119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442119 1 g 1g
16430858 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442119 Supplier Medchem Express Supplier No. HYB0276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ethionamide(2-ethylthioisonicotinamide) is an antibiotic used in the treatment of tuberculosis.

Ethionamide(2-ethylthioisonicotinamide) is an antibiotic used in the treatment of tuberculosis.

Specifications

Chemical Name or Material Ethionamide
Physical Form Powder
CAS 536-33-4
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H10N2S
For Use With (Application) COVID-19-immunoregulation
Synonym 2-Ethylthioisonicotinamide
Solubility Information DMSO : 250 mg/mL (1503.85 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (4.03 mM; Need ultrasonic)
SMILES S=C(C1=CC(CC)=NC=C1)N
Molecular Weight (g/mol) 166.24
Formula Weight 166.24
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.