missing translation for 'onlineSavingsMsg'
Learn More

Ethopabate, MedChemExpress

Product Code. 16478846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478846 100 mg 100mg
16432306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478846 Supplier Medchem Express Supplier No. HYB2138
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Ethopabate is an antiprotozoal agent which has been widely used to treat and prevent coccidiosis in chickens.

Ethopabate is an antiprotozoal agent which has been widely used to treat and prevent coccidiosis in chickens.

Specifications

Chemical Name or Material Ethopabate
Physical Form Solid
CAS 59-06-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15NO4
For Use With (Application) COVID-19-immunoregulation
Synonym Ethyl pabate
Solubility Information DMSO : 50 mg/mL (210.75 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=C(NC(C)=O)C=C1OCC
Molecular Weight (g/mol) 237.25
Formula Weight 237.25
Percent Purity 99.22%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.