missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethoxyquin

Product Code. 16650832 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 16650832

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-53-2 | C14H19NO | 217.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-53-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.31
InChI Key DECIPOUIJURFOJ-UHFFFAOYSA-N
IUPAC Name 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline
SMILES CCOC1=CC=C2NC(C)(C)C=C(C)C2=C1

Specifications

Boiling Point 161.0°C (1.6 kPa)
Flash Point 137°C
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in methanol, acetone, chloroform,, ethanol, toluene and hexane
Formula Weight 217.31
Chemical Name or Material Ethoxyquin
compliance-icons
Product Identifier
  • Ethoxyquin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ethoxyquin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.