missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%, Thermo Scientific Chemicals

Product Code. p-7055483
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471109 1 g 1g
15481109 5 g 5g
15491109 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471109 Supplier Thermo Scientific Alfa Aesar Supplier No. H64073.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51145-57-4 | Molecular Formula: C9H15NO3 | Molecular Weight: 185.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51145-57-4
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD00087563
InChI Key LQSOVGAUOHMPLK-SOFGYWHQSA-N
PubChem CID 2763877
IUPAC Name ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate
SMILES CCOC(=O)C(=CN(C)C)C(=O)C

Specifications

Color Red
Quantity 1 g
Synonym ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate
Formula Weight 185.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl 2-acetyl-3-(dimethylamino)acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.