missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromophenylacetate, 98%

Product Code. 11430150
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430150 250 mg 250mg
11440150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430150 Supplier Thermo Scientific Alfa Aesar Supplier No. H50630.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2178-24-7 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2178-24-7
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00051921
InChI Key MZQXAVZPEZUJIJ-UHFFFAOYSA-N
Synonym ethyl 2-2-bromophenyl acetate, ethyl 2-bromophenylacetate, ethyl 2-bromophenyl acetate, benzeneacetic acid, 2-bromo-, ethyl ester, acmc-20al1y, 2-bromo-phenyl-acetic acid ethyl ester, ethyl o-bromophenylacetate, ethyl-2-bromophenylacetate, ethyl 2-bromo-phenylacetate, ethyl 2-2-bromophenyl-acetate
PubChem CID 2780092
IUPAC Name ethyl 2-(2-bromophenyl)acetate
SMILES CCOC(=O)CC1=CC=CC=C1Br

Specifications

Melting Point 35°C to 38°C
Quantity 250 mg
Formula Weight 243.1
Percent Purity 98%
Chemical Name or Material Ethyl 2-bromophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.