missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(ethoxycarbonylmethoxy)benzoate, 97%, Thermo Scientific™

Product Code. 11313057
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313057 50 g 50g
11303057 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313057 Supplier Thermo Scientific Alfa Aesar Supplier No. L13430.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56424-77-2
Molecular Formula C13H16O5
Molecular Weight (g/mol) 252.266
MDL Number MFCD00151820
InChI Key NIUVYKIVCCMXLR-UHFFFAOYSA-N
Synonym ethyl 2-2-ethoxy-2-oxoethoxy benzoate, ethyl 2-ethoxycarbonylmethoxy benzoate, ethyl o-carboethoxymethylsalicylate, ethyl 2-2-ethoxy-2-oxo-ethoxy benzoate, ethyl 2-2-ethoxycarbonyl phenoxy acetate, 2-2-ethoxy-2-oxoethoxy benzoic acid ethyl ester, ethyl 2-2-ethoxy-2-oxidanylidene-ethoxy benzoate, benzoic acid,2-2-ethoxy-2-oxoethoxy-, ethyl ester
PubChem CID 347810
IUPAC Name ethyl 2-(2-ethoxy-2-oxoethoxy)benzoate
SMILES CCOC(=O)COC1=CC=CC=C1C(=O)OCC

Specifications

Boiling Point 180°C (4 mmHg)
Refractive Index 1.514
Quantity 50 g
Beilstein 2659411
Formula Weight 252.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-(ethoxycarbonylmethoxy)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.