missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-iodobenzoate, 98%

Product Code. 11391997
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391997 5 g 5g
11302007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391997 Supplier Thermo Scientific Alfa Aesar Supplier No. L07650.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1829-28-3 | C9H9IO2 | 276.07 g/mol

Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1829-28-3
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.07
MDL Number MFCD00051796
InChI Key QOUFDDUDXYJWHV-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid ethyl ester, rarechem al bi 0562, ethyl iodobenzoate, ethyl-2-iodobenzoate, ethyl 2-iodanylbenzoate, acmc-1ag98, 2-ethoxycarbonyl iodobenzene, benzoic acid, 2-iodo-,ethyl ester, benzoic acid, 2-iodo-, ethyl ester
PubChem CID 2829347
IUPAC Name ethyl 2-iodobenzoate
SMILES CCOC(=O)C1=CC=CC=C1I

Specifications

Boiling Point 163°C to 165°C (23 mmHg)
Refractive Index 1.584
Quantity 5 g
Beilstein 2084297
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 276.08
Percent Purity 98%
Chemical Name or Material Ethyl 2-iodobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.