missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitrobenzoate, 97%

Product Code. 11460431
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460431 25 g 25g
11470431 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460431 Supplier Thermo Scientific Alfa Aesar Supplier No. A19994.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-34-4 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-34-4
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00014696
InChI Key CPNMAYYYYSWTIV-UHFFFAOYSA-N
Synonym ethyl o-nitrobenzoate, ethyl nitrobenzoate, benzoic acid, o-nitro-, ethyl ester, benzoic acid, 2-nitro-, ethyl ester, nitrobenzoesaureathylester, acmc-1aul6, 2-nitro-benzoic acid ethyl ester
PubChem CID 69123
IUPAC Name ethyl 2-nitrobenzoate
SMILES CCOC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 26°C to 30°C
Boiling Point 172°C to 174°C (18 mmHg)
Quantity 25 g
Beilstein 2050729
Merck Index 14,3832
Formula Weight 195.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-nitrobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.