missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-octynoate, 98%

Product Code. 11408657
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11408657 25 g 25g
11498647 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11408657

Brand: Thermo Scientific Alfa Aesar A19784.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10519-20-7 | C10H16O2 | 168.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10519-20-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00015286
InChI Key QPMDWYXUSMRVKT-UHFFFAOYSA-N
Synonym ethyl 2-octynoate, 2-octynoic acid, ethyl ester, 2-octynoic acid ethyl ester, ethyl 2-octynate, ethyl heptynecarbonate, unii-xth780u0n3, ethyl heptine carbonate, ethyl n-oct-2-ynoate, ethyl heptyne carboxylate, 2-octynoic acid, ethylester
PubChem CID 66338
IUPAC Name ethyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OCC

Specifications

Density 0.91
Boiling Point 85°C (6 mmHg)
Flash Point 99°C (210°F)
Linear Formula CH3(CH2)4C≡CCO2CH2CH3
Refractive Index 1.447
Quantity 25 g
Beilstein 1759822
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material Ethyl 2-octynoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.