missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(trifluoromethyl)benzoate, 98%, Thermo Scientific™

Product Code. 11493307
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493307 100 g 100g
11473307 5 g 5g
11483307 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493307 Supplier Thermo Scientific Alfa Aesar Supplier No. A16215.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-62-8
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD00013556
InChI Key BQLMZZVRHPZRBQ-UHFFFAOYSA-N
Synonym ethyl 2-trifluoromethyl benzoate, 2-trifluoromethyl benzoic acid ethyl ester, 2-trifluoromethylbenzoic acid, ethyl ester, acetic acid,2,2,2-trifluoro-, 2-ethylphenyl ester, acmc-20al1u, ethyl-2-trifluoromethyl benzoate, ethyl 2-trifluoromethyl benzoate #, 2-trifluoromethylbenzoic acid ethyl ester, 2-trifluoromethyl-benzoic acid ethyl ester
PubChem CID 521815
IUPAC Name ethyl 2-(trifluoromethyl)benzoate
SMILES CCOC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Density 1.459
Boiling Point 90°C to 91°C (10 mmHg)
Refractive Index 1.45
Quantity 100 g
Beilstein 3290053
Formula Weight 218.18
Percent Purity 98%
Chemical Name or Material Ethyl 2-(trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.