missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(1,3-thiazol-2-yl)benzoate, 97%, Thermo Scientific™

Product Code. 10377594
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377594 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10377594 Supplier Thermo Scientific Maybridge Supplier No. CC36523DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 886851-29-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H11NO2S
MDL Number MFCD09064945
Quantity 1 g
Synonym ethyl 3-1,3-thiazol-2-yl benzoate, benzoic acid,3-2-thiazolyl-, ethyl ester, ethyl 3-thiazol-2-yl benzoate, benzoic acid,3-2-thiazolyl-,ethyl ester
InChI Key LHYDRQQHIJUWDE-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=CC(=C1)C2=NC=CS2
IUPAC Name ethyl 3-(1,3-thiazol-2-yl)benzoate
Molecular Weight (g/mol) 233.285
PubChem CID 24229479
Formula Weight 233.29
Percent Purity 97%
Chemical Name or Material Ethyl 3-(1, 3-thiazol-2-yl)benzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.