missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-bromophenyl)propionate, 95%

Product Code. 15458188
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458188 25 g 25g
15448188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458188 Supplier Thermo Scientific Alfa Aesar Supplier No. H61633.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40640-98-0 | C11H13BrO2 | 257.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40640-98-0
Molecular Formula C11H13BrO2
Molecular Weight (g/mol) 257.13
MDL Number MFCD07772961
InChI Key STMHGPFYLQOGJD-UHFFFAOYSA-N
PubChem CID 12731927
IUPAC Name ethyl 3-(4-bromophenyl)propanoate
SMILES CCOC(=O)CCC1=CC=C(Br)C=C1

Spécification

Quantity 25 g
Synonym ethyl 3-4-bromophenyl propanoate, 3-4-bromophenyl propionic acid ethyl ester, benzenepropanoic acid, 4-bromo-, ethyl ester, ethyl 3-4-bromophenyl-propionate, 3-4-bromo-phenyl-propionic acid ethyl ester, ksc497s3n, ethyl3-4-bromophenyl propanoate, 4-bromobenzenepropanoic acid ethyl ester, 3-4-bromophenyl propanoic acid ethyl ester, 3-4-bromophenyl-propionic acid ethyl ester
Formula Weight 257.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 3-(4-bromophenyl)propionate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.