missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-bis(trifluoromethyl)benzoate, 97%, Thermo Scientific™

Product Code. 10275081 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10275081 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 10275081

Brand: Thermo Scientific Acros 326360050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 96617-71-9
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H8F6O2
Refractive Index 1.4145 to 1.4165
MDL Number MFCD01320684
Quantity 5 g
Synonym ethyl 3,5-bis trifluoromethyl benzoate, 3,5-bis-trifluoromethyl-benzoic acid ethyl ester, 3,5-bis trifluoromethyl benzoic acid ethyl ester, rarechem al bi 0207, acmc-20al07, ethyl 3?5-bis trifluoromethyl benzoate, ethyl 3 5-bis trifluoromethyl benzoate, benzoic acid, 3,5-bis trifluoromethyl-, ethyl ester
InChI Key NBWZJMOEFTYMOF-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
IUPAC Name ethyl 3,5-bis(trifluoromethyl)benzoate
Molecular Weight (g/mol) 286.17
PubChem CID 2733458
Formula Weight 286.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 3, 5-bis(trifluoromethyl)benzoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3,5-bis(trifluoromethyl)benzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.