missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-dichloro-4-hydroxybenzoate, 99%, Thermo Scientific™

Product Code. 11408950
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408950 5 g 5g
11474114 25 g 25g
11484114 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408950 Supplier Thermo Scientific Alfa Aesar Supplier No. A12489.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 3,5-dichloro-4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17302-82-8
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
MDL Number MFCD00016420
InChI Key WMKNGSJJEMFQOT-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester
PubChem CID 28460
IUPAC Name ethyl 3,5-dichloro-4-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl

Specifications

Melting Point 110°C to 112°C
Quantity 5 g
Beilstein 2649965
Solubility Information Soluble in methanol.
Formula Weight 235.07
Percent Purity 99%
Chemical Name or Material Ethyl 3,5-dichloro-4-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.