missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate, 97%, Thermo Scientific™

Product Code. 11336855
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11336855 5 g Amber Glass Bottle 5g
10205374 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336855 Supplier Thermo Scientific Maybridge Supplier No. CC62623DE
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-99-6
Molecular Formula C14H14N2O3
Molecular Weight (g/mol) 258.28
MDL Number MFCD09702407
InChI Key ALDBUDYDJSLBJO-UHFFFAOYSA-N
Synonym ethyl 3-6-methylpyrazin-2-yl oxy benzoate, ethyl 3-6-methylpyrazin-2-yloxy benzoate, benzoic acid,3-6-methyl-2-pyrazinyl oxy-,ethyl ester
PubChem CID 24229683
IUPAC Name ethyl 3-(6-methylpyrazin-2-yl)oxybenzoate
SMILES CCOC(=O)C1=CC(OC2=NC(C)=CN=C2)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 258.28
Percent Purity 97%
Chemical Name or Material Ethyl 3-[(6-methylpyrazin-2-yl)oxy]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.