missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxybenzoate, 99%

Product Code. p-7022521
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11458256

Brand: Thermo Scientific Alfa Aesar A15343.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7781-98-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002296
InChI Key MWSMNBYIEBRXAL-UHFFFAOYSA-N
Synonym ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate
PubChem CID 24522
IUPAC Name ethyl 3-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)O

Specifications

Melting Point 72°C to 76°C
Boiling Point 187°C to 188°C (31 mmHg)
Quantity 25 g
Beilstein 2208566
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 3-hydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.