missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenoxypropionate, 98%

Product Code. 11351977
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11351977 10 g 10g
11361977 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11351977 Supplier Thermo Scientific Alfa Aesar Supplier No. L07606.09
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22409-91-2 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22409-91-2
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00026919
InChI Key TXKJOPXGYSFUNC-UHFFFAOYSA-N
PubChem CID 347973
IUPAC Name ethyl 3-phenoxypropanoate
SMILES CCOC(=O)CCOC1=CC=CC=C1

Specifications

Melting Point 22°C to 24°C
Density 1.084
Boiling Point 141°C to 143°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5005
Quantity 10 g
Beilstein 2331072
Synonym ethyl 3-phenoxypropionate, 3-phenoxypropanoic acid ethyl ester, acmc-20akz9, propanoic acid,3-phenoxy-, ethyl ester, propanoic acid, 3-phenoxy-, ethyl ester
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material Ethyl 3-phenoxypropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.