missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzene Sulfonamide Carbamate, TRC

Product Code. 30011575
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011575 100 mg 100mg
30135634 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011575 Supplier Toronto Research Chemicals Supplier No. TRCE901960100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14511-59-2
Molecular Formula C19 H21 Cl N2 O6 S
Molecular Weight (g/mol) 440.9
IUPAC Name ethyl N-[4-[2-[(5-chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonylcarbamate
SMILES CCOC(=O)NS(=O)(=O)c1ccc(CCNC(=O)c2cc(Cl)ccc2OC)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H21ClN2O6S/c1-3-28-19(24)22-29(25,26)15-7-4-13(5-8-15)10-11-21-18(23)16-12-14(20)6-9-17(16)27-2/h4-9,12H,3,10-11H2,1-2H3,(H,21,23)(H,22,24)
Formula Weight 440.0809
Chemical Name or Material Ethyl 4-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzene sulfonamide carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.