missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4-dimethyl-3-oxovalerate, 97%

Product Code. 11478627
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478627 25 g 25g
11488627 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478627 Supplier Thermo Scientific Alfa Aesar Supplier No. A19752.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17094-34-7 | C9H16O3 | 172.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17094-34-7
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.22
MDL Number MFCD00042886
InChI Key VUYNTIDSHCJIKF-UHFFFAOYSA-N
Synonym ethyl pivaloylacetate, ethyl 4,4,4-trimethylacetoacetate, ethyl 4,4-dimethyl-3-oxovalerate, 4,4-dimethyl-3-oxovaleric acid ethyl ester, pivaloylacetic acid ethyl ester, ethyl 4,4-dimethyl-3-oxo-pentanoate, pentanoic acid, 4,4-dimethyl-3-oxo-, ethyl ester, 4,4-dimethyl-3-oxo-pentanoic acid ethyl ester, ethylpivaloylacetate, pubchem19816
PubChem CID 86950
IUPAC Name ethyl 4,4-dimethyl-3-oxopentanoate
SMILES CCOC(=O)CC(=O)C(C)(C)C

Specifications

Density 0.967
Boiling Point 88°C to 90°C (10 mmHg)
Quantity 25 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl 4,4-dimethyl-3-oxovalerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.