missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Acetamidobenzoate, TRC

Product Code. 30133802
Change view
Click to view available options
Quantity:
500 mg
25 g
Unit Size:
25g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133802 25 g 25g
30012273 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133802 Supplier Toronto Research Chemicals Supplier No. TRCE89470025G
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5338-44-3
Molecular Formula C11 H13 N O3
Molecular Weight (g/mol) 207.23
Synonym N-Acetylbenzocaine, Benzoic acid, 4-(acetylamino)-, ethyl ester, Benzoic acid, p-acetamido-, ethyl ester (6CI,8CI), 4'-Carbethoxyacetanilide, Ethyl 4-acetamidobenzoate, Ethyl 4-acetylaminobenzoate, Ethyl N-acetyl-4-aminobenzoate, N-(4-(Ethoxycarbonyl)phenyl)acetamide, NSC 28987, NSC 3160
IUPAC Name ethyl 4-acetamidobenzoate
SMILES CCOC(=O)c1ccc(NC(=O)C)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C11H13NO3/c1-3-15-11(14)9-4-6-10(7-5-9)12-8(2)13/h4-7H,3H2,1-2H3,(H,12,13)
Formula Weight 207.0895
Chemical Name or Material N-Acetylbenzocaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.