missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromobenzoate, 98+%

Product Code. 11460173
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460173 250 g 250g
11440173 10 g 10g
11450173 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460173 Supplier Thermo Scientific Alfa Aesar Supplier No. A11365.30
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol

Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-bromobenzoate undergoes reduction with potassium diisobutyl-t-butoxyaluminum hydride (PDBBA) at 0°C to yield aldehydes. Reaction of ethyl 4-bromobenzoate and substituted benzyl chloride with zinc dust and a Pd catalyst is reported.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5798-75-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016329
InChI Key XZIAFENWXIQIKR-UHFFFAOYSA-N
Synonym 4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate
PubChem CID 22043
IUPAC Name ethyl 4-bromobenzoate
SMILES CCOC(=O)C1=CC=C(Br)C=C1

Specifications

Density 1.403
Boiling Point 262°C to 264°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 250 g
Beilstein 1867129
Merck Index 14,1408
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-bromobenzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.