missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-bromobenzoate, 99%

Product Code. 10387891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387891 50 mL Glass bottle 50mL
10096183 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387891 Supplier Thermo Scientific Acros Supplier No. 249760500
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5798-75-4 | C9H9BrO2 | 229.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 5798-75-4
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00016329
InChI Key XZIAFENWXIQIKR-UHFFFAOYSA-N
Synonym 4-bromobenzoic acid ethyl ester, ethyl p-bromobenzoate, benzoic acid, 4-bromo-, ethyl ester, ethyl4-bromobenzoate, ethyl-4-bromobenzoate, p-ethoxycarbonyl phenyl bromide, benzoic acid, p-bromo-, ethyl ester, 4-bromo-benzoic acid ethyl ester, 4-bromobenzoic acid, ethyl ester, ethyl-p-bromobenzoate
PubChem CID 22043
IUPAC Name ethyl 4-bromobenzoate
SMILES CCOC(=O)C1=CC=C(Br)C=C1

Especificaciones

Melting Point 18°C
Density 1.4300g/mL
Boiling Point 262°C to 264°C
Flash Point >110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CO2CH2CH3
Refractive Index 1.5440 to 1.547
Quantity 50 mL
Specific Gravity 1.43
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in polar solvents,insoluble in non-polar solvents
Formula Weight 229.06
Percent Purity 99%
Chemical Name or Material Ethyl 4-bromobenzoate, 99%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.