missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(bromomethyl)benzoate, 96%

Product Code. 11384086
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11384086

Brand: Thermo Scientific Alfa Aesar L19219.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26496-94-6 | C10H11BrO2 | 243.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26496-94-6
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.10
MDL Number MFCD03791307
InChI Key TWQLMAJROCNXEA-UHFFFAOYSA-N
Synonym ethyl 4-bromomethyl benzoate, 4-bromomethyl benzoic acid ethyl ester, 4-bromomethyl-benzoic acid ethyl ester, 4-ethoxycarbonylbenzyl bromide, benzoic acid, 4-bromomethyl-, ethyl ester, zlchem 925, acmc-1cknv, 4-carboethoxy-benzylbromide, 4-carboethoxybenzyl bromide, ethyl 4-bromomethyl-benzoate
PubChem CID 954261
IUPAC Name ethyl 4-(bromomethyl)benzoate
SMILES CCOC(=O)C1=CC=C(CBr)C=C1

Specifications

Melting Point 28°C to 33°C
Boiling Point 145°C (5 mmHg)
Refractive Index 1.5494
Quantity 5 g
UN Number UN1759
Beilstein 1869569
Formula Weight 243.1
Percent Purity 96%
Chemical Name or Material Ethyl 4-(bromomethyl)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.