missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxybenzoate, 99%

Product Code. 11484564
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484564 5000 g 5000g
11474564 250 g 250g
11464564 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484564 Supplier Thermo Scientific Alfa Aesar Supplier No. A13172.0I
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 120-47-8 | C9H10O3 | 166.176 g/mol

Ethyl 4-hydroxybenzoate (ethylparaben) was used to study the in-situderivatisation solid-phase microextraction procedure for determining parabens, related chlorophenols and triclosan in water. It is a novel sorbent for solid-phase extraction, was used to study its retention property. It has high extraction efficiency towards the compounds tested owing to the electrostatic interaction, hydrophobic interaction and hydrogen bonding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxybenzoate (ethylparaben) was used to study the in-situderivatisation solid-phase microextraction procedure for determining parabens, related chlorophenols and triclosan in water. It is a novel sorbent for solid-phase extraction, was used to study its retention property. It has high extraction efficiency towards the compounds tested owing to the electrostatic interaction, hydrophobic interaction and hydrogen bonding.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 116°C to 118°C
Boiling Point 297°C to 298°C (decomposition)
Quantity 5000 g
Beilstein 1101972
Merck Index 14,3837
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.