missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxybenzoate, 99%

Product Code. p-3737945 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Unit Size:
100g
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10317420

Brand: Thermo Scientific Acros 150251000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 120-47-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-47-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002353
InChI Key NUVBSKCKDOMJSU-UHFFFAOYSA-N
Synonym ethylparaben, ethyl paraben, ethyl p-hydroxybenzoate, mycocten, ethyl parasept, 4-hydroxybenzoic acid ethyl ester, easeptol, ethyl butex, ethyl parahydroxybenzoate, tegosept e
PubChem CID 8434
ChEBI CHEBI:86616
IUPAC Name ethyl 4-hydroxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 115°C to 118°C
Color White
Boiling Point 297°C to 298°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4CO2C2H5
Quantity 100 g
Beilstein 10, 159
Merck Index 15, 3890
Solubility Information Solubility in water: 1.7g/L (20°C). Other solubilities: freely soluble in alcohol and ether,soluble in carbon disulfide,slightly soluble in petroleum ether,chloroform
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-hydroxybenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-hydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.