missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-hydroxymandelate, 98%

Product Code. 11367435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367435 5 g 5g
11357435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367435 Supplier Thermo Scientific Alfa Aesar Supplier No. L05413.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68758-68-9 | C10H12O4 | 196.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68758-68-9
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00020180
InChI Key VLOUFSKXRCPIQR-UHFFFAOYNA-N
Synonym ethyl 2-hydroxy-2-4-hydroxyphenyl acetate, ethyl 4-hydroxymandelate, ethyl 4-hydroxy-dl-mandelate, ethyl alpha,4-dihydroxyphenylacetate, 4-hydroxymandelic acid ethyl ester, benzeneacetic acid, .alpha.,4-dihydroxy-, ethyl ester, benzeneacetic acid, alpha,4-dihydroxy-, ethyl ester, ethyl hydroxy 4-hydroxyphenyl acetate, ethyl hydroxy 4-hydroxyphenyl acetate #, 4-2-ethoxy-1-hydroxy-2-oxoethyl phenol
PubChem CID 111357
IUPAC Name ethyl 2-hydroxy-2-(4-hydroxyphenyl)acetate
SMILES CCOC(=O)C(O)C1=CC=C(O)C=C1

Specifications

Melting Point 128°C to 132°C
Quantity 5 g
Beilstein 3132065
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Ethyl 4-hydroxymandelate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.