missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methoxybenzoate, 98+%

Product Code. 11427876
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427876 50 g 50g
11437876 250 g 250g
11417876 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427876 Supplier Thermo Scientific Alfa Aesar Supplier No. A14008.18
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-30-4 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00015151
InChI Key FHUODBDRWMIBQP-UHFFFAOYSA-N
Synonym ethyl p-anisate, ethyl anisate, ethyl p-methoxybenzoate, p-anisic acid, ethyl ester, benzoic acid, 4-methoxy-, ethyl ester, 4-methoxybenzoic acid ethyl ester, benzoic acid, p-methoxy-, ethyl ester, ethyl p-anisoate, unii-kj95h2s7nm, p-anisic acid ethyl ester
PubChem CID 60979
IUPAC Name ethyl 4-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)OC

Specifications

Melting Point 7°C to 8°C
Density 1.111
Boiling Point 263°C
Flash Point 138°C (280°F)
Refractive Index 1.526
Quantity 50 g
Beilstein 2209700
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.