missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methoxyphenylacetate, 97%

Product Code. 10526641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10526641 50 g Glass bottle 50g
10235861 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10526641 Supplier Thermo Scientific Supplier No. 189840500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14062-18-1 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-18-1
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00040760
InChI Key DOCCDOCIYYDLGJ-UHFFFAOYSA-N
Synonym ethyl 2-4-methoxyphenyl acetate, ethyl 4-methoxyphenylacetate, 4-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-methoxy-, ethyl ester, ethyl p-methoxyphenylacetate, ethyl 4-methoxyphenyl acetate, 4-methoxy-phenyl-acetic acid ethyl ester, acmc-1bnx2, homoanisic acid ethyl ester
PubChem CID 84174
IUPAC Name ethyl 2-(4-methoxyphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)OC

Specifications

Density 1.0970g/mL
Boiling Point 108°C to 111°C (3.0 mmHg)
Flash Point 46°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CO2C2H5
Refractive Index 1.5065 to 1.5085
Quantity 50 g
Beilstein 10, I, 83
Specific Gravity 1.097
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material Ethyl 4-methoxyphenylacetate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-methoxyphenylacetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.