missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromo-2-chlorobenzoate, 97%, Thermo Scientific™

Product Code. 10646081 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
Packaging:
Glass Bottle
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 10646081

Brand: Thermo Scientific Acros 250030100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 76008-73-6
Color Yellow
Density 1.4800g/mL
Boiling Point 128.0°C to 130.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C9H8BrClO2
Refractive Index 1.5575 to 1.5595
MDL Number MFCD00013635
Quantity 10 mL
Specific Gravity 1.48
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
Molecular Weight (g/mol) 263.515
PubChem CID 144749
Formula Weight 263.52
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 5-bromo-2-chlorobenzoate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 5-bromo-2-chlorobenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.